missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphtheria Toxoid Antibody (17.6B9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23596-0.1ML
This item is not returnable.
View return policy
Description
Diphtheria Toxoid Monoclonal specifically detects Diphtheria Toxoid in Bacteria samples. It is validated for ELISA.
Specifications
| Diphtheria Toxoid | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100-1:2000 | |
| C. Diphtheria toxin, Corynebacterium diphtheriae toxin, Corynebacterium diphtheriae toxoid, DIP0222, Diptheria toxin, Diptheriae toxin, DTx | |
| Diphtheria toxoid prepared from toxin purified from C. diphtheriae subsp. intermedius Park-Williams no. 8 by formaldehyde inactivation (2), and adsorbed to aluminum hydroxide gel. | |
| 0.1 mL | |
| It reacts with diphtheria toxoid and also neutralizes diphtheria toxin as determined in a VERO cell assay | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
| ELISA | |
| 17.6B9 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction