missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphtheria Toxoid Antibody (17.6B9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 481.00
Specifications
| Antigen | Diphtheria Toxoid |
|---|---|
| Clone | 17.6B9 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
Description
Diphtheria Toxoid Monoclonal specifically detects Diphtheria Toxoid in Bacteria samples. It is validated for ELISA.Specifications
| Diphtheria Toxoid | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
| Diphtheria toxoid prepared from toxin purified from C. diphtheriae subsp. intermedius Park-Williams no. 8 by formaldehyde inactivation (2), and adsorbed to aluminum hydroxide gel. | |
| Primary | |
| Store at 4C. Do not freeze. |
| 17.6B9 | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Bacteria | |
| C. Diphtheria toxin, Corynebacterium diphtheriae toxin, Corynebacterium diphtheriae toxoid, DIP0222, Diptheria toxin, Diptheriae toxin, DTx | |
| IgG1 κ | |
| Protein A or G purified | |
| It reacts with diphtheria toxoid and also neutralizes diphtheria toxin as determined in a VERO cell assay |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title