missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylalanine methyl ester hydrochloride, 98%

Product Code. 11405093
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11405093

Brand: Thermo Scientific Alfa Aesar A12758.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5619-07-8 | C10H14ClNO2 | 215.68 g/mol

DL-Phenylalanine methyl ester hydrochloride, is an aminoacid which is widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Phenylalanine methyl ester hydrochloride, is an aminoacid which is widely used in pharmaceuticals and foods.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5619-07-8
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00066113
InChI Key SWVMLNPDTIFDDY-UHFFFAOYNA-N
Synonym dl-phenylalanine methyl ester hydrochloride, methyl 2-amino-3-phenylpropanoate hydrochloride, h-dl-phe-ome.hcl, h-l-phe-ome hcl, phenylalanine methyl ester hydrochloride, l-phenylalanine methylester hydrochloride, l-methyl phenylalanate hydrochloride, s-phenylalanine methyl ester hydrochloride, acmc-209oxm, acmc-1bga3
PubChem CID 3084017
IUPAC Name methyl 2-amino-3-phenylpropanoate;hydrochloride
SMILES Cl.COC(=O)C(N)CC1=CC=CC=C1

Specifications

Melting Point ∼160°C (decomposition)
Beilstein 3597947
Solubility Information Slightly soluble in water.
Quantity 25 g
Formula Weight 215.68
Percent Purity 98%
Chemical Name or Material DL-Phenylalanine methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.