missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA polymerase delta p50 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38341-20ul
This item is not returnable.
View return policy
Description
DNA polymerase delta p50 Polyclonal antibody specifically detects DNA polymerase delta p50 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| DNA polymerase delta p50 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| delta 2, regulatory subunit (50kD), polymerase (DNA directed), delta 2, regulatory subunit 50kDa | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-360 of human DNA polymerase delta p50 (NP_006221.2).,, Sequence:, LQPEEKCCVVGTLFKAMPLQPSILREVSEEHNLLPQPPRSKYIHPDDELVLEDELQRIKLKGTIDVSKLVTGTVLAVFGSVRDDGKFLVEDYCFADLAPQKPAPPLDTDRFVLLVSGLGLGGGGGESLLGTQLLVDVVTGQLGDEGEQCSAAHVSRVILAGNLLSHSTQSRDSINKAKYLTKKTQAASVEAVKMLDEILLQLSASVPVDVMPGEFDPTNYTLPQQPLHPCMFPLATAYSTL | |
| 20 μL | |
| Cell Cycle and Replication | |
| 5425 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction