missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA polymerase delta p50 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | DNA polymerase delta p50 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229962
|
Novus Biologicals
NBP3-38341-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230457
|
Novus Biologicals
NBP3-38341-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA polymerase delta p50 Polyclonal antibody specifically detects DNA polymerase delta p50 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| DNA polymerase delta p50 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 5425 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| delta 2, regulatory subunit (50kD), polymerase (DNA directed), delta 2, regulatory subunit 50kDa | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-360 of human DNA polymerase delta p50 (NP_006221.2).,, Sequence:, LQPEEKCCVVGTLFKAMPLQPSILREVSEEHNLLPQPPRSKYIHPDDELVLEDELQRIKLKGTIDVSKLVTGTVLAVFGSVRDDGKFLVEDYCFADLAPQKPAPPLDTDRFVLLVSGLGLGGGGGESLLGTQLLVDVVTGQLGDEGEQCSAAHVSRVILAGNLLSHSTQSRDSINKAKYLTKKTQAASVEAVKMLDEILLQLSASVPVDVMPGEFDPTNYTLPQQPLHPCMFPLATAYSTL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title