missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA polymerase sigma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56356SS
This item is not returnable.
View return policy
Description
DNA polymerase sigma Polyclonal specifically detects DNA polymerase sigma in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| DNA polymerase sigma | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500 | |
| NP_008930.1 | |
| TENT4A | |
| This antibody was generated by immunizing rabbits with a synthetic peptide containing amino acids 447-462 (GTASLKAVHHMSSPAI) of human TRF4-1, GenBank no NP_008930.1. | |
| 0.025 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 0.5% BSA with 0.02% Sodium Azide | |
| DNA polymerase kappa, DNA polymerase sigma, EC 2.7.7.7, LAK-1TUTase 5, PAP associated domain containing 7, POLKTRF41, POLSTUTASE5, polymerase (DNA directed) sigma, polymerase (DNA-directed) sigma, Terminal uridylyltransferase 5, Topoisomerase-related function protein 4-1, TRF4-1LAK1, TRF4PAP-associated domain-containing protein 7 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 11044 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction