missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA polymerase sigma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 213.00 - € 361.00
Specifications
| Antigen | DNA polymerase sigma |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18029893
|
Novus Biologicals
NB100-56356SS |
0.025 mL |
€ 213.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18701324
|
Novus Biologicals
NB100-56356 |
0.1 mL |
€ 361.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA polymerase sigma Polyclonal specifically detects DNA polymerase sigma in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| DNA polymerase sigma | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| NP_008930.1 | |
| 11044 | |
| This antibody was generated by immunizing rabbits with a synthetic peptide containing amino acids 447-462 (GTASLKAVHHMSSPAI) of human TRF4-1, GenBank no NP_008930.1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.5% BSA with 0.02% Sodium Azide | |
| DNA polymerase kappa, DNA polymerase sigma, EC 2.7.7.7, LAK-1TUTase 5, PAP associated domain containing 7, POLKTRF41, POLSTUTASE5, polymerase (DNA directed) sigma, polymerase (DNA-directed) sigma, Terminal uridylyltransferase 5, Topoisomerase-related function protein 4-1, TRF4-1LAK1, TRF4PAP-associated domain-containing protein 7 | |
| TENT4A | |
| IgG | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title