missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPEP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | DPEP2 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604348
|
Novus Biologicals
NBP3-00197-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633707
|
Novus Biologicals
NBP3-00197-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DPEP2 Polyclonal antibody specifically detects DPEP2 in Human samples. It is validated for ELISASpecifications
| DPEP2 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Endocrinology | |
| 0.2 um filtered solution in PBS | |
| 64174 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| dipeptidase 2, EC 3.4.13.19, MBD2 | |
| Produced in rabbits immunized with purified, recombinant Human DPEP2 (Accession#: AAH24021.1; Met1-Ser376) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title