Learn More
Sanplatec™ Dry Keeper Non-Auto
Product Code. 30245634
Click to view available options
Model:
C-3B
C-3BS
Unit Size:
Each
Product Code. 30245634
Brand: Sanplatec™ 0030
Only null left
Add to basket
Add to basket
A non-auto type desiccator excellent for use in dehumidification applications.
  • With a convenient pan for silica gel
  • Shelf plates can be adjusted in 8 steps at 28 mm intervals to match the size of stored items
  • Horizontal design for added stability
  • Inside humidity is slightly different depending on the set room temperature and usage conditions
  • Body cabinet of PVC, frame of ABS
  • White frame, transparent sides
Specifications
Cabinet Style Front-loading
Material PVC, ABS
Product Type Desiccator
Includes Silicagel tray, 3pcs of shelves
Model C-3B