Learn More
Sanplatec™ Dry Keeper Non-Auto
Click to view available options
Model:
C-3B
C-3BS
Unit Size:
Each
Description
- With a convenient pan for silica gel
- Shelf plates can be adjusted in 8 steps at 28 mm intervals to match the size of stored items
- Horizontal design for added stability
- Inside humidity is slightly different depending on the set room temperature and usage conditions
- Body cabinet of PVC, frame of ABS
- White frame, transparent sides
Specifications
Specifications
| Cabinet Style | Front-loading |
| Material | PVC, ABS |
| Product Type | Desiccator |
| Includes | Silicagel tray, 3pcs of shelves |
| Model | C-3B |