missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DSC1 Recombinant Rabbit Monoclonal Antibody (23GB5210), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553865
This item is not returnable.
View return policy
Description
The desmosomal cadherin desmocollin DSC1 is expressed in upper epidermis where strong adhesion is required. DSC1 is a type I membrane protein (4) required for strong adhesion and barrier maintenance in epidermis and contributes to epidermal differentiation.The DSC1 gene comprises 17 exons spanning approximately 33 kb on 18q12.1.
Specifications
| DSC1 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P55849, Q08554 | |
| DSC1 | |
| A synthesized peptide derived from human Desmocollin 1 (824-894AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5210 | |
| Unconjugated | |
| DSC1 | |
| 1110020A10Rik; AI507491; Cadherin family member 1; CDHF1; desmocollin 1; Desmocollin1; desmocollin-1; Desmosomal glycoprotein 2/3; DG2/DG3; DSC1; Dsc1a; Dsc1b | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13505, 1823, 291759 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction