missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DSC1 Recombinant Rabbit Monoclonal Antibody (23GB5210), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 207.00 - € 562.00
Specifications
| Antigen | DSC1 |
|---|---|
| Clone | 23GB5210 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089237
![]() |
Invitrogen™
MA553864 |
100 μL |
€ 562.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087597
|
Invitrogen™
MA553865 |
20 μL |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DSC1 Recombinant Monoclonal antibody specifically detects DSC1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| DSC1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P55849, Q08554 | |
| 13505, 1823, 291759 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5210 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110020A10Rik; AI507491; Cadherin family member 1; CDHF1; desmocollin 1; Desmocollin1; desmocollin-1; Desmosomal glycoprotein 2/3; DG2/DG3; DSC1; Dsc1a; Dsc1b | |
| A synthesized peptide derived from human Desmocollin 1 (824-894AA). | |
| Primary | |
| -20°C | |
| DSC1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title