missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DTX3L Monoclonal Antibody (K24022_3F7), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556192

 View more versions of this product

Product Code. 30210948

  • € 868.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: RVRRKLESYF QSSKSSGGGE CTVSTQEHEA PGTFRVEFSE RAAKERVLKK GEHQILVDEK PVPIFLVPTE NSIKKNTRPQ ISSLTQSQAE TPSGDMHQHE

Component of the shelterin complex (telosome) that is involved in the regulation of telomere length and protection. Shelterin associates with arrays of double-stranded TTAGGG repeats added by telomerase and protects chromosome ends. Without its protective activity, telomeres are no longer hidden from the DNA damage surveillance and chromosome ends are inappropriately processed by DNA repair pathways. Promotes binding of POT1 to single-stranded telomeric DNA. Modulates the inhibitory effects of POT1 on telomere elongation. The ACD-POT1 heterodimer enhances telomere elongation by recruiting telomerase to telomeres and increasing its processivity. May play a role in organogenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DTX3L
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q8TDB6
DTX3L
Recombinant human DTX3L (immunogen range is 21-120).
200 μL
Primary
Human
Antibody
IgG1
Immunoprecipitation, Western Blot
K24022_3F7
Unconjugated
DTX3L
AU042200; Bbap; BC023741; B-lymphoma- and BAL-associated protein; deltex 3 like, E3 ubiquitin ligase; deltex 3-like; deltex 3-like (Drosophila); deltex E3 ubiquitin ligase 3L; DTX3L; E3 ubiquitin-protein ligase DTX3L; Protein deltex-3-like; RGD1565144; rhysin 2; Rhysin2; rhysin-2; RING-type E3 ubiquitin transferase DTX3L
Mouse
Protein A
RUO
151636
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.