missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DTX3L Monoclonal Antibody (K24022_3F7), Invitrogen™
Mouse Monoclonal Antibody
€ 480.00 - € 846.00
Specifications
| Antigen | DTX3L |
|---|---|
| Clone | K24022_3F7 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30210948
|
Invitrogen™
MA556192 |
200 μL |
€ 846.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30213468
|
Invitrogen™
MA555812 |
50 μL |
€ 480.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: RVRRKLESYF QSSKSSGGGE CTVSTQEHEA PGTFRVEFSE RAAKERVLKK GEHQILVDEK PVPIFLVPTE NSIKKNTRPQ ISSLTQSQAE TPSGDMHQHE
DTX3L Monoclonal antibody specifically detects DTX3L in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| DTX3L | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AU042200; Bbap; BC023741; B-lymphoma- and BAL-associated protein; deltex 3 like, E3 ubiquitin ligase; deltex 3-like; deltex 3-like (Drosophila); deltex E3 ubiquitin ligase 3L; DTX3L; E3 ubiquitin-protein ligase DTX3L; Protein deltex-3-like; RGD1565144; rhysin 2; Rhysin2; rhysin-2; RING-type E3 ubiquitin transferase DTX3L | |
| Recombinant human DTX3L (immunogen range is 21-120). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| DTX3L |
| K24022_3F7 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8TDB6 | |
| 151636 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title