Learn More
DWK Life Sciences DURAN™ Glass Stopper, Clear, Standard Taper Ground Joint, Octagonal flat head
Clear DURAN™ glass ground stopper.
Brand:  DWK Life Sciences 216240701
89.78 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15425924
  • € 94.50 / Pack of 10
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
Description
Description
Semi-hollow or solid DURAN™ glass stoppers can be used to close glassware items such as flasks, separatory funnels or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
  • All DURAN™ glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to Standard Taper ground joint
  • Octagonal shaped head
Specifications
Specifications
Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders
Boro 3.3
No
10/Pk.
ST 19/26