Learn More
DWK Life Sciences DURAN™ Stopcock Key, hollow, threaded glass handle, ground, with three part retaining device
To be used for higher tightness requirements.
Brand: DWK Life Sciences 286041803
926.25 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 5.00000kg
Description
Hollow DURAN™ tap plugs are well suited for work with temperature changes and higher tightness requirements.
- Hollow tap plug with thread, glass handle and 3-piece tap safety device
Specifications
1.5 mm | |
Yes | |
50/Pk. |
Borosilicate Glass (3.3), Polypropylene | |
12.5 NS | |
When working with temperature changes and higher tightness requirements |