Learn More
DYNC1LI1 Monoclonal Antibody (OTI1H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501511
Encompass_Preferred
 View more versions of this product
Product Code. 17554441
  • € 540.87 / 100µL
Description
Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules. May play a role in binding dynein to membranous organelles or chromosomes. Probably involved in the microtubule-dependent transport of pericentrin. Is required for progress throuh the spindle assembly checkpoint. The phosphorylated form appears to be involved in the selective removal of MAD1L1 and MAD1L2 but not BUB1B from kinetochores.
Specifications
Specifications
DYNC1LI1
Monoclonal
0.4 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9Y6G9
DYNC1LI1
Full length human recombit protein of human DYNC1LI1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1H4
Unconjugated
DYNC1LI1
DLC-A, DNCLI1, LIC1
Mouse
Affinity Chromatography
RUO
51143
-20° C, Avoid Freeze/Thaw Cycles