missing translation for 'onlineSavingsMsg'
Learn More

DYNC1LI1 Monoclonal Antibody (OTI1H4), TrueMAB™, OriGene

Mouse Monoclonal Antibody

POA

Specifications

Antigen DYNC1LI1
Clone OTI1H4
Concentration 0.4 mg/mL
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17574441
Encompass_Preferred
View Documents
Origene Technologies
TA501511S
30 μL
POA
30µL
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17554441
Encompass_Preferred
View Documents
Origene Technologies
TA501511
100 μL
POA
100µL
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

DYNC1LI1 Monoclonal antibody specifically detects DYNC1LI1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications

Specifications

DYNC1LI1
0.4 mg/mL
Unconjugated
Mouse
Human
51143
Primary
DYNC1LI1
OTI1H4
Monoclonal
Liquid
RUO
Q9Y6G9
IgG2b
-20° C, Avoid Freeze/Thaw Cycles
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.