missing translation for 'onlineSavingsMsg'
Learn More

(E)-Metaflumizone, TRC

Product Code. 30471912
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471912 100 mg 100mg
30483524 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30471912 Supplier Toronto Research Chemicals Supplier No. TRCM225826100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 852403-68-0
Molecular Formula C24 H16 F6 N4 O2
Molecular Weight (g/mol) 506.4
Synonym (2E)-2-[2-(4-Cyanophenyl)-1-[3-(trifluoromethyl)phenyl]ethylidene]-N-[4-(trifluoromethoxy)phenyl]hydrazinecarboxamide, (E)-Metaflumizone
IUPAC Name 1-[(E)-[2-(4-cyanophenyl)-1-[3-(trifluoromethyl)phenyl]ethylidene]amino]-3-[4-(trifluoromethoxy)phenyl]urea
SMILES FC(F)(F)Oc1ccc(NC(=O)N\N=C(/Cc2ccc(cc2)C#N)\c3cccc(c3)C(F)(F)F)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H16F6N4O2/c25-23(26,27)18-3-1-2-17(13-18)21(12-15-4-6-16(14-31)7-5-15)33-34-22(35)32-19-8-10-20(11-9-19)36-24(28,29)30/h1-11,13H,12H2,(H2,32,34,35)/b33-21+
Formula Weight 506.1177
Chemical Name or Material (E)-Metaflumizone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.