Learn More
EAAT2/GLT1 Antibody (28F9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26322-50ul
This item is not returnable.
View return policy
Description
EAAT2/GLT1 Monoclonal antibody specifically detects EAAT2/GLT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| EAAT2/GLT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 28F9 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
| GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2 | |
| A synthesized peptide derived from Human EAAT2/GLT1 [UniProt P43004] | |
| 50 μL | |
| Hypoxia, Neuroscience, Neurotransmission | |
| 6506 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |