missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAAT2/GLT1 Antibody (28F9), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 355.00 - € 568.00
Specifications
| Antigen | EAAT2/GLT1 |
|---|---|
| Clone | 28F9 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30037015
|
Novus Biologicals
NBP3-26322-50ul |
50 μL |
€ 355.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30033073
|
Novus Biologicals
NBP3-26322-100ul |
100 μL |
€ 568.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EAAT2/GLT1 Monoclonal antibody specifically detects EAAT2/GLT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
| EAAT2/GLT1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2 | |
| A synthesized peptide derived from Human EAAT2/GLT1 [UniProt P43004] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 28F9 | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Hypoxia, Neuroscience, Neurotransmission | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 6506 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title