Learn More
Ansell™ Easy Flex 47-200 Series Nitrile Medium Weight Gloves
Shop All Ansell Products
Click to view available options
Size:
6.5
7
8
9
10
Unit Size:
12 pairs
144 pairs
Description
- Much better resistance to snags, puncture and abrasion.
- Nitrile coating repels oil and grease
- Safely handle smooth objects like boxes or appliances with ease
- Lasts five times longer than an ordinary cotton glove
- Cut-and-sewn construction
- Interlock cotton liner
- 3/4 Dipped
- Knitwrist cuff
Specifications
Specifications
| Back Material | Nitrile |
| Certifications/Compliance | CE Mark, ANSI, CATEGORY II EN 388 : 2111 |
| Cuff Material | Nitrile |
| Coating Material | Nitrile |
| Color | Green |
| Exterior Finish | No Texture |
| Hand | Specific |
| Latex Free | Yes |
| Material | Cotton |
| Product Type | Mediumweight Gloves |
| Show More |