missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Antibody (5P10D3), Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-03943
This item is not returnable.
View return policy
Description
EBI3 Monoclonal specifically detects EBI3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| IL-27/IL-35 EBI3 Subunit | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 0.5-5 ug/ml, Flow Cytometry 1-3ug/10^6cells, ELISA reported in scientific literature (PMID 30660991) | |
| cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 5P10D3 | |
| Unconjugated | |
| O35228 | |
| EBI3 | |
| Full-length recombinant mouse EBI3 protein | |
| 0.1 mg | |
| Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| 10148 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction