missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Antibody (5P10D3), Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
€ 226.00 - € 509.00
Specifications
| Antigen | IL-27/IL-35 EBI3 Subunit |
|---|---|
| Clone | 5P10D3 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5-5 ug/ml, Flow Cytometry 1-3ug/10^6cells, ELISA reported in scientific literature (PMID 30660991) |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18020357
|
Novus Biologicals
NBP2-03943SS |
0.025 mg |
€ 226.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18077556
|
Novus Biologicals
NBP2-03943 |
0.1 mg |
€ 509.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EBI3 Monoclonal specifically detects EBI3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| IL-27/IL-35 EBI3 Subunit | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Rat | |
| Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| O35228 | |
| 10148 | |
| Full-length recombinant mouse EBI3 protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 5P10D3 | |
| Western Blot 0.5-5 ug/ml, Flow Cytometry 1-3ug/10^6cells, ELISA reported in scientific literature (PMID 30660991) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
| EBI3 | |
| IgG2b κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title