Learn More
EBV Early Antigens Antibody (1108-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-34333-0.02mg
 View more versions of this product
Product Code. 18473242
  • € 290.00 / 0.02mg
Description
EBV Early Antigens Monoclonal antibody specifically detects EBV Early Antigens in Virus samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EBV Early Antigens
Monoclonal
0.2 mg/mL
Immunocytochemistry/ Immunofluorescence 1-2 ug/mL
Chronic ulcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, p40, p51, p63, p73L, TP63, Transformation-related protein 63, Tumor protein 63, Tumor protein p73-like
Affinity Purified early antigen polypeptides from induced Raji cells precipitated with African Burkitt's lymphoma sera
0.02 mg
Primary
Store at 4C.
IgG1 κ
Immunocytochemistry
1108-1
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Virus