missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBV Early Antigens Antibody (1108-1), Novus Biologicals™
Mouse Monoclonal Antibody
€ 274.05 - € 480.90
Specifications
| Antigen | EBV Early Antigens |
|---|---|
| Clone | 1108-1 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunocytochemistry/ Immunofluorescence 1-2 ug/mL |
| Applications | Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18424572
|
Novus Biologicals
NBP2-34333-0.1mg |
0.1 mg |
€ 509.00 € 480.90 / 0.10mg Save € 28.10 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18473242
|
Novus Biologicals
NBP2-34333-0.02mg |
0.02 mg |
€ 290.00 € 274.05 / 0.02mg Save € 15.95 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EBV Early Antigens Monoclonal antibody specifically detects EBV Early Antigens in Virus samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| EBV Early Antigens | |
| 0.2 mg/mL | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Chronic ulcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, p40, p51, p63, p73L, TP63, Transformation-related protein 63, Tumor protein 63, Tumor protein p73-like | |
| IgG1 κ | |
| Protein A or G purified |
| 1108-1 | |
| Immunocytochemistry/ Immunofluorescence 1-2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Affinity Purified early antigen polypeptides from induced Raji cells precipitated with African Burkitt's lymphoma sera | |
| Primary | |
| Store at 4C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title