Learn More
ECHS1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-11305-20UL
Description
The ECHS1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ECHS1. This antibody recognizes human, mouse, rat antigen. The ECHS1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| ECHS1 | |
| 1000 μg/mL | |
| ECHS1, Enoyl CoA hydratase 1, SCEH | |
| 20 μL | |
| 1892 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |