Learn More
ECHS1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-11305-20UL
 View more versions of this product
Product Code. 18028025
  • € 145.00 / 20µL
Description
The ECHS1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ECHS1. This antibody recognizes human, mouse, rat antigen. The ECHS1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
ECHS1
1000 μg/mL
ECHS1, Enoyl CoA hydratase 1, SCEH
20 μL
1892
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa