missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECHS1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | ECHS1 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18028025
|
Proteintech
CL488-11305-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18089795
|
Proteintech
CL488-11305-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
The ECHS1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ECHS1. This antibody recognizes human, mouse, rat antigen. The ECHS1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Spezifikation
| ECHS1 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| ECHS1, Enoyl CoA hydratase 1, SCEH | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 1892 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts