Learn More
Ansell EDGE™ 48-702 Protective Gloves
Product Code. 18829912 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
144 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
18829912 11 144 pairs
18889902 7 144 pairs
18899902 8 144 pairs
18809912 9 144 pairs
18819912 10 144 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 18829912 Supplier Ansell Supplier No. 48702110
Only null left
Add to basket
Add to basket
Medium-duty safety gloves with a firm grip, ensuring greater cut and abrasion protection.
  • Made of a seamless liner, palm dipped into micro-foam nitrile coating.
  • The micro-foam nitrile coating provides excellent tactile sensitivity and dexterity vs. foam nitrile coating.
  • The micro-foam nitrile coating provides a good grip and excellent abrasion resistance, compatible with slight oil.
  • Very comfortable to wear, the breathable coating keeps hands cool and dry.
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Nitrile Foam
Color Black
Cut Resistance Rating Level B
Hand Specific
Latex Free Yes
Material HPPE, Nylon, Polyester, Spandex, Foam Nitrile
Product Type Industrial Gloves
Size 11