missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEA1 Antibody (SR1246), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21679-100ul
This item is not returnable.
View return policy
Description
EEA1 Monoclonal antibody specifically detects EEA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| EEA1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1246 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2 | |
| A synthesized peptide derived from human EEA1 (Uniprot #: Q15075) | |
| 100 μg | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Endosome Markers, Immunology, Innate Immunity, Lysosome Markers, Membrane Trafficking and Chaperones | |
| 8411 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction