missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEA1 Antibody (SR1246), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | EEA1 |
|---|---|
| Clone | SR1246 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619673
|
Novus Biologicals
NBP3-21679-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651794
|
Novus Biologicals
NBP3-21679-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EEA1 Monoclonal antibody specifically detects EEA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| EEA1 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2 | |
| A synthesized peptide derived from human EEA1 (Uniprot #: Q15075) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1246 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Endosome Markers, Immunology, Innate Immunity, Lysosome Markers, Membrane Trafficking and Chaperones | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 8411 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title