missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1E1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 624.00
Specifications
| Antigen | EEF1E1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18229804
|
Novus Biologicals
NBP2-55640 |
100 μL |
€ 624.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641469
|
Novus Biologicals
NBP2-55640-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EEF1E1 Polyclonal specifically detects EEF1E1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| EEF1E1 | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 9521 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GLHRFIVDLTVQEKEKYLNVSRWFCHIQHYPGIRQHLSSVVFIKNRLYTNSH | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| AIMP3Aminoacyl tRNA synthetase complex-interacting multifunctional protein 3, ARS-interacting multifunctional protein 3, Elongation factor p18, eukaryotic translation elongation factor 1 epsilon 1, eukaryotic translation elongation factor 1 epsilon-1, Multisynthase complex auxiliary component p18, P18, p18 component of aminoacyl-tRNA synthetase complex | |
| EEF1E1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title