missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24504
This item is not returnable.
View return policy
Description
eIF4B Polyclonal specifically detects eIF4B in Human, Mouse, Primate samples. It is validated for Western Blot.
Specifications
| eIF4B | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| EIF-4B, eukaryotic translation initiation factor 4B, PRO1843 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, chimpanzee, mouse, horse, opossum and Xenopus. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.005-1.0 ug/ml | |
| P23588 | |
| EIF4B | |
| A portion of amino acids 570-611 of human eIF4B was used as the immunogen. | |
| 0.1 mg | |
| Core ESC Like Genes, mTOR Pathway, Stem Cell Markers | |
| 1975 | |
| Human, Mouse, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction