missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 199.00 - € 351.00
Specifications
| Antigen | eIF4B |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.005-1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18109131
|
Novus Biologicals
NBP2-24504SS |
0.025 mg |
€ 199.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18128601
|
Novus Biologicals
NBP2-24504 |
0.1 mg |
€ 351.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF4B Polyclonal specifically detects eIF4B in Human, Mouse, Primate samples. It is validated for Western Blot.Specifications
| eIF4B | |
| Western Blot 0.005-1.0 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, mTOR Pathway, Stem Cell Markers | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| EIF-4B, eukaryotic translation initiation factor 4B, PRO1843 | |
| EIF4B | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, chimpanzee, mouse, horse, opossum and Xenopus. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Primate | |
| P23588 | |
| 1975 | |
| A portion of amino acids 570-611 of human eIF4B was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title