missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4B Antibody (S07-2H2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19859-50ul
This item is not returnable.
View return policy
Description
eIF4B Monoclonal antibody specifically detects eIF4B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| eIF4B | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin | |
| EIF-4B, eukaryotic translation initiation factor 4B, PRO1843 | |
| A synthetic peptide of human eIF4B (Uniprot # P23588) | |
| 50 μg | |
| Core ESC Like Genes, mTOR Pathway, Phospho Specific, Stem Cell Markers | |
| 1975 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S07-2H2 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction