missing translation for 'onlineSavingsMsg'
Learn More

eIF4B Antibody (S07-2H2), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 421.00 - € 645.00

Specifications

Antigen eIF4B
Clone S07-2H2
Concentration 0.3 mg/mL
Dilution Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18664793
View Documents
Novus Biologicals
NBP3-19859-100ul
100 μg
€ 645.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18624683
View Documents
Novus Biologicals
NBP3-19859-50ul
50 μg
€ 421.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

eIF4B Monoclonal antibody specifically detects eIF4B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

eIF4B
0.3 mg/mL
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Core ESC Like Genes, mTOR Pathway, Phospho Specific, Stem Cell Markers
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
1975
IgG
Affinity purified
S07-2H2
Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human
EIF-4B, eukaryotic translation initiation factor 4B, PRO1843
A synthetic peptide of human eIF4B (Uniprot # P23588)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.