missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF5 Antibody (6D6G6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61763-0.025ml
This item is not returnable.
View return policy
Description
EIF5 Monoclonal specifically detects EIF5 in Human, Monkey samples. It is validated for Western Blot, ELISA.
Specifications
| EIF5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| eIF-5, EIF-5A, eukaryotic translation initiation factor 5 | |
| Mouse | |
| 49.2 kDa | |
| 0.025 mL | |
| 1983 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 6D6G6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| EIF5 | |
| Purified recombinant fragment of human EIF5 (AA: 1-300) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction