missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF5 Antibody (6D6G6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 552.00
Specifications
| Antigen | EIF5 |
|---|---|
| Clone | 6D6G6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18210524
|
Novus Biologicals
NBP2-61763 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698768
|
Novus Biologicals
NBP2-61763-0.025ml |
0.025 mL |
€ 203.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EIF5 Monoclonal specifically detects EIF5 in Human, Monkey samples. It is validated for Western Blot, ELISA.Specifications
| EIF5 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Human, Monkey | |
| 1983 | |
| Purified recombinant fragment of human EIF5 (AA: 1-300) expressed in E. Coli. | |
| Primary | |
| 49.2 kDa |
| 6D6G6 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Unconjugated | |
| Mouse | |
| eIF-5, EIF-5A, eukaryotic translation initiation factor 5 | |
| EIF5 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title