Learn More
Elma™ Lab Clean A20sf Ultrasonic Cleaning Solution
Ideal for cleaning calibrated and volumetric labware such as pipets and burets. Elma™ Lab Clean A20sf Cleaning Solution removes light grease, lime soap residues, fingerprints and dust.
Brand:  Elma™ 8000055
Additional Details : Weight : 1.12000kg
 View more versions of this product
Product Code. 10389834
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alert:
Inhalation of spray may be harmful and may cause respiratory irritation. May be harmful if swallowed.
Description
Description
  • Use in ultrasonic bath or labware washer
  • Mildly alkaline, contains ammonia
  • Surfactant-free
  • Residue-free - Leaves no chemical residue after rinsing and drying
  • Enables undisturbed draining and clear reading in glass vessels
  • Dilute to 1% by volume for use in an ultrasonic cleaner or to 0.5% in labware washers
  • Can also be used as soaking bath (>2%) without ultrasonics
  • Biodegradable
  • Available in 10 and 25L containers
Applications
  • Cleans glass, stainless steel, ceramics, and plastics
Note
  • Not for aluminum, magnesium and alloys of light metals
Specifications
Specifications
Ultrasonic Cleaning Product
Mildly alkaline
Cleaning Pipets and Other Volumetric Labware in Ultrasonic Cleaners and Labware Washers
1 L
Liquid
Keep above 32°F, shelf life 4 years
20 Pk.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • elma lab clean A20sf
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sodium carbonate, ammonia, tetrapotassium pyrophosphate