missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ELOVL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30900-25ug
This item is not returnable.
View return policy
Description
ELOVL4 Polyclonal antibody specifically detects ELOVL4 in Human samples. It is validated for Western Blot
Specifications
| ELOVL4 | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL | |
| ADMD, cancer/testis antigen 118, CT118, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 4, elongation of very long chain fatty acids protein 4, FLJ17667, FLJ92876,3-keto acyl-CoA synthase ELOVL4, Stargardt disease 3 (autosomal dominant), STGD2, STGD3 | |
| Synthetic peptdide derived from the human ELOVL4 protein | |
| 25 μg | |
| Lipid and Metabolism, Vision | |
| 6785 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction