missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ELOVL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 213.00 - € 508.00
Specifications
| Antigen | ELOVL4 |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066888
|
Novus Biologicals™
NBP3-30900-25ug |
25 μg |
€ 213.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067090
|
Novus Biologicals™
NBP3-30900-100ug |
100 μg |
€ 508.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELOVL4 Polyclonal antibody specifically detects ELOVL4 in Human samples. It is validated for Western BlotSpecifications
| ELOVL4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Vision | |
| Phosphate buffered saline | |
| 6785 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1-5 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ADMD, cancer/testis antigen 118, CT118, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 4, elongation of very long chain fatty acids protein 4, FLJ17667, FLJ92876,3-keto acyl-CoA synthase ELOVL4, Stargardt disease 3 (autosomal dominant), STGD2, STGD3 | |
| Synthetic peptdide derived from the human ELOVL4 protein | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title