Learn More
Invitrogen™ EMC4 Recombinant Rabbit Monoclonal Antibody (23GB5315), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552839
This item is not returnable.
View return policy
Description
May mediate anti-apoptotic activity.
Specifications
| EMC4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5J8M3, Q9CZX9 | |
| EMC4 | |
| A synthesized peptide derived from human EMC4 (22-65AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5315 | |
| Unconjugated | |
| EMC4 | |
| 2610318K02Rik; Cell proliferation-inducing gene 17 protein; EMC4; ER membrane protein complex subunit 4; HSPC184; PIG17; TMEM85; Transmembrane protein 85 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 296049, 51234, 68032 | |
| -20°C | |
| Liquid |