missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EMC4 Recombinant Rabbit Monoclonal Antibody (23GB5315), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 195.00 - € 468.00
Specifications
| Antigen | EMC4 |
|---|---|
| Clone | 23GB5315 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088476
![]() |
Invitrogen™
MA552838 |
100 μL |
€ 468.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088524
|
Invitrogen™
MA552839 |
20 μL |
€ 195.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EMC4 Recombinant Monoclonal antibody specifically detects EMC4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| EMC4 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q5J8M3, Q9CZX9 | |
| 296049, 51234, 68032 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5315 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2610318K02Rik; Cell proliferation-inducing gene 17 protein; EMC4; ER membrane protein complex subunit 4; HSPC184; PIG17; TMEM85; Transmembrane protein 85 | |
| A synthesized peptide derived from human EMC4 (22-65AA). | |
| Primary | |
| -20°C | |
| EMC4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title