missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emerin Antibody (8F5G2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52460
This item is not returnable.
View return policy
Description
Emerin Monoclonal specifically detects Emerin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications
| Emerin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, CyTOF-ready | |
| emerin, Emery-Dreifuss muscular dystrophy, LEM domain containing 5, STAEDMDLEMD5 | |
| Mouse | |
| 29 kDa | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| 8F5G2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| EMD | |
| Purified recombinant fragment of human *** (AA: 1-222) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| 2010 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction