missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emerin Antibody (8F5G2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 552.00
Specifications
| Antigen | Emerin |
|---|---|
| Clone | 8F5G2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18299810
|
Novus Biologicals
NBP2-52460 |
0.1 mg |
€ 552.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643337
|
Novus Biologicals
NBP2-52460-0.025mg |
0.025 mg |
€ 203.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Emerin Monoclonal specifically detects Emerin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
| Emerin | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| emerin, Emery-Dreifuss muscular dystrophy, LEM domain containing 5, STAEDMDLEMD5 | |
| EMD | |
| IgG1 | |
| Protein A or G purified |
| 8F5G2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 2010 | |
| Purified recombinant fragment of human *** (AA: 1-222) expressed in E. Coli. | |
| Primary | |
| 29 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title