Learn More
Emerin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand:  Novus Biologicals NBP1-87692
 View more versions of this product
Product Code. 18709763
  • € 557.00 / 0.10mL
Description
Emerin Polyclonal specifically detects Emerin in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Emerin
emerin, Emery-Dreifuss muscular dystrophy, LEM domain containing 5, STAEDMDLEMD5
Affinity Purified
IgG
Western Blot
EMD
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.