missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emerin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
€ 353.00
Specifications
| Antigen | Emerin |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18474162
|
Novus Biologicals
NBP1-87692-25ul |
25 μL |
€ 353.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18709763
|
Novus Biologicals
NBP1-87692 |
N/A
|
N/A | ||||||
Description
Emerin Polyclonal specifically detects Emerin in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Emerin | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat, Hamster | |
| P50402 | |
| 2010 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ASSYSFSDLNSTRGDADMYDLPKKEDALLYQSKGYNDDYYEESYFTTRTYGEPESAGPSRAVRQSVTSFPDADAFHHQVHDDDLLSSSEEECKDRERPMYGRDSACQSITHY | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| emerin, Emery-Dreifuss muscular dystrophy, LEM domain containing 5, STAEDMDLEMD5 | |
| EMD | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title