missing translation for 'onlineSavingsMsg'
Learn More

Eperisone Hydrochloride, TRC

Product Code. 17961735
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17961735 100 mg 100mg
17934635 1 g 1g
17923335 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17961735 Supplier Toronto Research Chemicals Supplier No. TRCE565800100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56839-43-1
Molecular Formula C17 H25 N O . Cl H
Molecular Weight (g/mol) 295.85
Synonym 1-Propanone, 1-(4-ethylphenyl)-2-methyl-3-(1-piperidinyl)-, hydrochloride (1:1), 1-Propanone, 1-(4-ethylphenyl)-2-methyl-3-(1-piperidinyl)-, hydrochloride (9CI), (+/-)-Eperisone hydrochloride, 4'-Ethyl-2-methyl-3-piperidinopropiophenone hydrochloride, E 0646, EMPP, Eperisone hydrochloride, Mional, Myonal, Myosone plus, Rapisone, Skelact
IUPAC Name 1-(4-ethylphenyl)-2-methyl-3-piperidin-1-ylpropan-1-one;hydrochloride
SMILES Cl.CCc1ccc(cc1)C(=O)C(C)CN2CCCCC2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H25NO.ClH/c1-3-15-7-9-16(10-8-15)17(19)14(2)13-18-11-5-4-6-12-18;/h7-10,14H,3-6,11-13H2,1-2H3;1H
Formula Weight 295.1703
Percent Purity >95%
Chemical Name or Material Eperisone Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.