missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB4 Antibody (5B5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47434-0.025ml
This item is not returnable.
View return policy
Description
EphB4 Monoclonal specifically detects EphB4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| EphB4 | |
| Monoclonal | |
| Unconjugated | |
| EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 | |
| Mouse | |
| 108 kDa | |
| 0.025 mL | |
| Protein Kinase | |
| 2050 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 5B5 | |
| Western Blot 1: 500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| EPHB4 | |
| Purified recombinant extracellular fragment of human EphB4 fused with hIgGFc tag expressed in HEK293 cell line. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction