missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB4 Antibody (5B5), Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00
Specifications
| Antigen | EphB4 |
|---|---|
| Clone | 5B5 |
| Dilution | Western Blot 1: 500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
EphB4 Monoclonal specifically detects EphB4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| EphB4 | |
| Western Blot 1: 500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 2050 | |
| Purified recombinant extracellular fragment of human EphB4 fused with hIgGFc tag expressed in HEK293 cell line. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 5B5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 | |
| EPHB4 | |
| IgG1 | |
| Unpurified | |
| 108 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title